Beakers, low form, borosilicate glass, heavy-duty, double scale, KIMAX®, KIMBLE
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP14005-2000 SP14005-1000
SCERSP14005-2000EA 732 EUR
SCERSP14005-2000 SCERSP14005-1000
Beakers, low form, borosilicate glass, heavy-duty, double scale, KIMAX®, KIMBLE
Bekers
Manufactured from 33 expansion, low extractable borosilicate glass. Thick uniform walls throughout and extra wall thickness built into the evenly tooled top rim.
  • Uniformity of construction allows for use on hot plates
  • All sizes have a durable matte finish marking area and a white graduated scale
  • Design meets ASTM specification E960, type II requirements
Heavy-duty glass 'thick wall' beakers offer superior mechanical strength and durability. Improved safety when used under extreme conditions such as mechanized washing and rough handling.
Certificaten: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.