Sharps containers, code YELLOW, SHARPAK™
Leverancier: Synergy Health
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SU02230 SU02430 SU02630 SU03530 SU06430
129-1052EA 18 EUR
129-1052 129-1055 129-1058 129-1060 VEROSU06430
Sharps containers, code YELLOW, SHARPAK™
Bakken Containers voor scherpe materialen
All SHARPAK™ Code YELLOW sharps containers feature yellow coloured closures and are recommended for the segregation, collection and disposal of partially discharged sharps waste which are NOT contaminated with cytotoxic or cytostatic medicines.
  • Yellow colour coded closures to meet the recommendations on waste segregation as detailed in the Department of Health's technical memorandum HTM-07-01
  • Secure closure with locking facility
  • Distinctive colouring and clear labelling
  • Sequential numbering for traceability
  • Supplied assembled
Code YELLOW SHARPAK™More Product Information