Chiral columns, ReproSil® Chiral-OM-R
Leverancier: DR MAISCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R65.OMR.S1546 R65.OMR.S0546 R65.OMR.S1046 R65.OMR.S2546 R60.OMR.S2546 R60.OMR.S1546 R60.OMR.S0546 R63.OMR.S1546 R63.OMR.S1046 R63.OMR.S0546
554-2634EA 0 EUR
554-2634 554-2632 554-2633 554-2635 554-2604 554-2603 554-2602 554-2617 554-2616 554-2615
Chiral columns, ReproSil® Chiral-OM-R
Kolommen voor chromatografie
USP L40 column: cellulose tris-3,5-dimethylphenylcarbamate coated to porous silica particles.