Semi-preparative and preparative silica material, ReproSil®-Pur
Leverancier: DR MAISCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R115.AQ.1000 R10.AQ.1000 R130.AQ.1000 RA10.9E.1000 RA115.9E.S1002 R15.8E.S1546
554-2583EA 0 EUR
554-2583 554-2574 554-2584 554-2636 MAISRA115.9E.S1002 MAISR15.8E.S1546
Semi-preparative and preparative silica material, ReproSil®-Pur
Sorbenten voor chromatografie Sorbenten voor HPLC
Semi-preparative and preparative spherical silica material allows the clean-up of raw products from mg up to kg amounts, depending on the column ID and length. Spherical materials are easy to pack in columns.
  • Spherical ultra-clean silica material
  • Base decativated
  • Hydrophilic endcapping
  • Mean pore size 120 Å
  • Specific surface 300 m²/g