Capillary columns, Velocity-WAX
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N9306314 N9306315 N9306322
568-0336EA 1130 EUR
568-0336 568-0337 568-0340
Capillary columns, Velocity-WAX
Kolommen voor chromatografie
This column is ideal for intermediate to high polarity compounds. General GC applications include essential oils, polar industrial chemicals and alcohol mixtures.
Polyethylene glycol.
Thermal stability: 20 to 250 °C
Suitable replacement for: DB-FFAP, HP-20M, SUPELCOWAX™ 10, CP-WAX 52CB, SUPEROX® II, CB-WAX, STABILWAX®, BP-20, Elite™-CW, 007-CW, Carbowax®, HP-Innowax.