Filter bags, BagFilter®
Leverancier: INTERSCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
111425 112425 113510 111625
432-0006EA 471 EUR
432-0006 432-0007 INSC113510 INSC111625
Filter bags, BagFilter®
Zakken Homogenisatiezakken
These lateral filter bags are with non-woven filter that are ideal for the homogenisation and automatic filtration of the samples.
  • Particle-free solution for easy reading of the colonies
  • Compatible with any blender
  • Sterile and approved for food contact
  • Multilayer® reinforced multicoated complex
BagFilter® P: Ideal for pipetting, filter porosity <250 microns.
BagFilter® S: Ideal for pouring, with a weld to retain particles, filter porosity <250 microns.
BagFilter Pull-Up®: Ideal for pipetting small volumes/PCR, filter porosity <50 microns.