Blender bags, BagLight®
Leverancier: INTERSCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
131025 132025 133025 132225 132525
INSC131025EA 147 EUR
INSC131025 INSC132025 INSC133025 INSC132225 INSC132525
Blender bags, BagLight®
Zakken Homogenisatiezakken
These non-filter sample preparation bags are ideal for homogenising samples.
  • Robust/ultra resistant
  • Resistant to freezing
  • Compatible with any blender
  • Sterile and approved for food contact
BagLight® PolySilk®: Made of PolySilk® polyolefin complex, flexible and transparent. Available for different volume.
BagLight® Multilayer®: Made of Multilayer® reinforced complex, reinforced weldings.
BagLight® PolySilk® HD: Made of high density PolySilk®. They are rigid, matte and slightly opaque.