Micro pipettes, disposable, Microcaps®
Leverancier: DRUMMOND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1-000-0300 1-000-0010-55 1-000-9000 1-000-0005 1-000-0040 1-000-0060 1-000-0070 1-000-0080 1-000-0150 1-000-0250 1-000-0400 1-000-0800 1-000-2000 1-000-0447 1-000-0005-64 1-000-00025 1-000-0033 1-000-0066 1-000-0350 1-000-0750 1-000-0090 1-000-0050-55
612-4864EA 44.1 EUR
612-4864 612-2470 612-4865 DRUM1-000-0005 DRUM1-000-0040 DRUM1-000-0060 DRUM1-000-0070 DRUM1-000-0080 612-4863 DRUM1-000-0250 DRUM1-000-0400 DRUM1-000-0800 DRUM1-000-2000 DRUM1-000-0447 612-4859 612-4858 DRUM1-000-0033 DRUM1-000-0066 DRUM1-000-0350 DRUM1-000-0750 DRUM1-000-0090 DRUM1-000-0050-55
Micro pipettes, disposable, Microcaps®
Pipetten Micro-capillairen
Micro pipettes are precision-bore glass capillary tubes cut to predetermined lengths so that fill volumes are constant and precise. Capillary action draws the fluid into the tube and a squeeze of the bulb dispenses it with accuracy of ±1%.
For fast, safe, clean transfer of liquids in micro quantities. Suitable for both general and hazardous microchemical applications.
Bestelinformatie: Delivery includes 100 pipettes and one bulb dispenser.