Inline check valves, standard 1/4-28, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CV-3301 CV-3302 CV-3315 CV-3316
UPCHCV-3301EA 182 EUR
UPCHCV-3301 UPCHCV-3302 554-3060 UPCHCV-3316
Inline check valves, standard 1/4-28, Upchurch Scientific®
Ventielen Terugslagventielen
These standard inline check valves are made from gold plated stainless steel.
  • The spring-actuated sealing system eliminates backflow, which prevents upstream contamination or damage
  • Cracking pressure of 3 psi (0,2 bar)
  • Excellent chemical resistance