VHP unions for capillary tubing, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
UH-432 UH-436 UH-632
554-3273EA 347 EUR
554-3273 UPCHUH-436 554-3275
VHP unions for capillary tubing, Upchurch Scientific®
Fittingen en connectoren
Unions with stainless steel bodies and PEEK fittings.
  • Pressure rated up to 15000 psi (1034 bar)
  • Options to direct-connect both ¹/₃₂" ext.Ø tubing and 360 µm ext.Ø tubing