Low pressure unions, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-602 P-603 P-620 P-622 P-623 P-630 P-631 P-645 P-702 P-702-01 P-703 P-703-01 P-708-02
554-3121EA 12.7 EUR
554-3121 554-3122 554-3123 554-3124 554-3125 UPCHP-630 UPCHP-631 UPCHP-645 554-3156 554-3157 554-3158 554-3159 UPCHP-708-02
Low pressure unions, Upchurch Scientific®
Fittingen en connectoren
These low pressure unions are made from PEEK, ETFE, Delrin or PP.
  • Pressure rating up to 1000 psi (69 bar) for PEEK union assemblies
  • Chemically resistant