Bulkhead unions, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-134 P-440 P-440-01 P-441 P-441-01 P-441-01BLK P-441-01N P-441BLK P-441G P-441N U-658
554-3092EA 39.5 EUR
554-3092 554-3100 554-3101 554-3102 554-3103 554-3105 554-3107 554-3109 554-3110 554-3111 UPCHU-658
Bulkhead unions, Upchurch Scientific®
Fittingen en connectoren
Designed for plumbing tubing through equipment housing. Each union has unique ³/₈-24 external threads and comes complete with a stainless steel nut and lock washer to hold it in place.
  • Requires a ³/₈" hole to mount
  • Recommended torque limit 15 in.– lbs (1,7 N-m)