VHP tees, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
UH-402 UH-427 UH-700 UH-750
UPCHUH-402EA 122 EUR
UPCHUH-402 554-3272 554-3276 UPCHUH-750
VHP tees, Upchurch Scientific®
Fittingen en connectoren
  • Pressure rated to 30000 psi (2070 bar) for tubing tees (¹/₁₆" Ø ext.)
  • Pressure rated to 15000 psi (1034 bar) for micro capillary tubing tees