High pressure micro capillary tees and crosses, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-775 P-775-01 P-777 P-777-01 P-885 P-888 P-888-01 P-890 P-890-01 P-891
554-3189EA 298 EUR
554-3189 554-3190 554-3191 554-3192 554-3208 554-3210 554-3211 554-3212 554-3213 UPCHP-891
High pressure micro capillary tees and crosses, Upchurch Scientific®
Fittingen en connectoren
PEEK.
  • Pressure rating up to 4000 psi (276 bar) for P-775, P-777
  • Pressure rating up to 5000 psi (345 bar) for P-885, P-888, P-890