High pressure speciality tees, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-540 M-540A U-433 U-466 U-466-02 U-466S
554-3083EA 477 EUR
554-3083 554-3084 UPCHU-433 554-3259 554-3260 554-3261
High pressure speciality tees, Upchurch Scientific®
Fittingen en connectoren
PEEK or stainless steel.
  • Pressure rating up to 5000 psi (345 bar)