Low pressure manifolds and Y connectors, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-150 P-150-01 P-155 P-190 P-190-01 P-191 P-512 P-512-01 P-513 P-514 P-514-01 P-515-01
554-3094EA 292 EUR
554-3094 554-3095 UPCHP-155 554-3098 554-3099 UPCHP-191 554-3117 554-3118 UPCHP-513 554-3119 554-3120 UPCHP-515-01
Low pressure manifolds and Y connectors, Upchurch Scientific®
Fittingen en connectoren
PEEK.
  • Pressure rating up to 1000 psi (69 bar) for manifolds and Y connectors for ¹/₁₆" ext. Ø tubing
  • Pressure rating up to 500 psi (34 bar) for manifolds and Y connectors for ¹/₈" ext. Ø tubing