English threaded adapters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-652 P-654 P-669-01 P-671
554-3137EA 38.7 EUR
554-3137 554-3138 554-3146 UPCHP-671
English threaded adapters, Upchurch Scientific®
Fittingen en connectoren
These adapters are made from PEEK or PTFE. Female to male 10-32 to 1/4-28 adapters.
  • Pressure rating up to 1000 psi (69 bar)