MicroTight® adapters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-770 P-881 P-881-01 P-882 P-882-01 UH-630
554-3183EA 94.6 EUR
554-3183 554-3204 554-3205 554-3206 554-3207 554-3274
MicroTight® adapters, Upchurch Scientific®
Fittingen en connectoren
These adapters are made from PEEK or stainless steel. They create a true zero dead volume (ZDV) connection between ¹/₁₆" ext. Ø tubing and capillary tubing.
  • Pressure rating up to 4000 psi (276 bar) for P-770
  • Pressure rating up to 5000 psi (345 bar) for P-881 and P-882
  • Pressure rating up to 12000 psi (827 bar) for UH-630