Barbed adapters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-639 P-646 P-647 P-648 P-663 P-664 P-665 P-668 P-689 P-691 P-692 D-647 D-648 D-646
UPCHP-639EA 212 EUR
UPCHP-639 554-3134 554-3135 UPCHP-648 554-3144 UPCHP-664 UPCHP-665 554-3145 554-3152 UPCHP-691 554-3153 554-3066 554-3067 554-3065
Barbed adapters, Upchurch Scientific®
Fittingen en connectoren
PP, ETFE or PEEK. The barbed inserts of the swivel barb adapters spin freely from the threaded nut in order to prevent the tubing from twisting during installation.