Barbed luer adapters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-850 P-851 P-852 P-853 P-855 P-856 P-857 P-858 P-859 P-870
UPCHP-850EA 4 EUR
UPCHP-850 UPCHP-851 UPCHP-852 UPCHP-853 UPCHP-855 UPCHP-856 UPCHP-857 UPCHP-858 UPCHP-859 UPCHP-870
Barbed luer adapters, Upchurch Scientific®
Fittingen en connectoren
PP or ETFE. Use these barbed adapters to connect peristaltic-type flexible tubing for general, low pressure applications, such as plumbing Ismatec® peristaltic pumps. Due to the low melting point of PP, these adapters are not autoclavable, however, they can be sterilised via gamma radiation. The ETFE connectors have superiour solvent resistance and a higher temperature rating (80 °C).
Order Now
Specificaties
Environmentally Preferable