Peristaltic tubing adapters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-757 P-767
554-3178EA 160 EUR
554-3178 UPCHP-767
Peristaltic tubing adapters, Upchurch Scientific®
Fittingen en connectoren
PEEK polymer adapters connect peristaltic tubing to standard rigid or semi-rigid tubing.
These unique adapters connect peristaltic tubing to standard ¹/₁₆" or ¹/₈" ext. Ø tubing. A specially-designed nose allows the peristaltic tubing to simply press fit over the nose and then be held tightly in place by the retainer sleeve. Tubing with ¹/₁₆" ext. Ø may then be connected with the flangeless fittings supplied with the adapter. To connect the peristaltic tubing to ¹/₈" ext. Ø tubing, simply replace the supplied fittings with the choice of flangeless fittings. One popular application forMore Product Information