Capillary columns, Biodiesel range
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0541180SG53 0541181SG53
568-0370EA 541 EUR
568-0370 568-0371
Capillary columns, Biodiesel range
Kolommen voor chromatografie
SGE has evaluated each ASTM and EN Biodiesel method to demonstrate the suitability of BPX-BIOD GC columns to achieve optimal separation.
Each BPX-BIOD GC column is tested at the maximum operating temperature for the column, and specific tests are undertaken to ensure the column meets inertness specifications and separation performance.