Semi-prep inline filters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A-410 A-411 OC-802 OC-805 A-330
554-3035EA 317 EUR
554-3035 UPCHA-411 554-3088 554-3090 554-3026
Semi-prep inline filters, Upchurch Scientific®
Filters Lijnfilters
Semi-prep inline filters
- Designed for high flow applications
- Flow rates up to 100 ml/min
- Pressure rated to 7500 psi (517 bar)
  • Designed for high flow applications
  • Flow rates up to 100 ml/min
  • Pressure rated to 7500 psi (517 bar)
Biocompatible semi-prep inline filters
- Stainless steel body, PEEK end fittings and PEEK frit
- Ideal for many higher flow analytical, semi-prep and preparative applications
- Pressure rated to 6000 psi (414 bar)