Low pressure tees and crosses, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-632 P-632-01 P-633 P-633-01 P-634 P-635 P-712 P-712-01 P-712-01R P-713 P-713-01 P-714 P-714-01 P-722 P-723
554-3130EA 80 EUR
554-3130 554-3131 554-3132 554-3133 UPCHP-634 UPCHP-635 554-3160 554-3161 554-3162 554-3163 554-3164 554-3165 554-3166 UPCHP-722 UPCHP-723
Low pressure tees and crosses, Upchurch Scientific®
Fittingen en connectoren
ETFE or PEEK.
  • Pressure rating up to 1000 psi (69 bar) for tees and crosses for ¹/₁₆" ext. Ø tubing
  • Pressure rating up to 500 psi (34 bar) for tees and crosses for ¹/₈" ext. Ø tubing