Adhesive film seals for microplates, SecureSeal™
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T329-3 T329-4
291-0900EA 979 EUR
291-0900 291-0032
Adhesive film seals for microplates, SecureSeal™
Afdichtingen Afdichtingen van microtiterplaten
Adhesive sealing films reduce sample-to-sample or well-to-well contamination and/or spill over. SecureSeal™ has the differential advantage of perforated end tabs and a multiple split backing which allow for easier and more accurate positioning and more secure sealing. The polyester based film with acrylic adhesive is inert and thus compatible with almost all microplate procedures. SecureSeal™ is less than 0,001% moisture permeable under high humidity conditions and less than 0,01% oxygen permeable.
  • Functional temperature range of −70 to +100 °C
  • DMSO resistant