French square bottles, clear, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W216885 W216883 W216884
WHEAW216885EA 159 EUR
WHEAW216885 WHEAW216883 WHEAW216884
French square bottles, clear, WHEATON®
Flessen Glazen flessen
French square bottles are manufactured from clear soda lime glass that conforms to USP Type III requirements. Design allows for full utilisation of any storage area, maximising space efficiency. Each case of bottles is packaged in convenience packs with caps attached to maintain cleanliness. A partitioned carton protects bottles from scratches during shipping, and they become useful for storing samples. Each case of bottles also includes a hazardous communication label and tamper evident seal if labelling and/or sealing is required. Extra screw caps can be purchased separately.
Bestelinformatie: Caps are supplied attached to bottles.