Jerricans, narrow neck, with built-in spout, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H109350000 H109360000 H109370000
BELAH109350000EA 174 EUR
BELAH109350000 BELAH109360000 BELAH109370000
Jerricans, narrow neck, with built-in spout, SP Bel-Art
Flessen Jerrycans
HDPE, translucent.
  • Good chemical resistance
  • Temperature resistant from −100 to +120 °C
  • Stable sturdy design with integral graduations in gallons and litres, leakproof closure
These jerricans have a unique 20 cm long spout with a built-in air vent for smooth pouring. The spout retracts into the threaded collar to conserve space, and the collar is sealed by a leak proof screw cap. An extra hand grip on the bottom gives positive control when pouring. The 5 gal. jerrican has a 2,5 cm int. Ø pouring spout; other sizes have 1,9 cm int. Ø spouts. Graduated in gallons and liters, they feature extra headspace for expansion of contents.