Wide opening crimp top vials, Target™ LoVial™ and ColorBand™
Leverancier: THERMO NATIONAL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C4011-95 C4011-88
NSCAC4011-95EA 259 EUR
NSCAC4011-95 NSCAC4011-88
Wide opening crimp top vials, Target™ LoVial™ and ColorBand™
flesjes
Vials feature a 40% larger opening than standard opening vials to reduce needle damage. They are manufactured from superior quality 33 expansion borosilicate clear glass Type 1 and fit most autosamplers that accommodate the 12×32 mm vial format.
Order Now
Specificaties
Environmentally Preferable