Ferrules for gas chromatography
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CR2111XX CR211200 CR213165 CR213200 CR213164
VARICR2111XXEA 76 EUR
VARICR2111XX VARICR211200 VARICR213165 VARICR213200 VARICR213164
Ferrules for gas chromatography
Fittingen en connectoren
Vespel/graphite and precision graphite ferrules for chromatography applications.
  • Does not require a back ferrule
Graphite ferrules are made from 100% high pure graphite with temperature limit of 450 °C. Ferrules are precision formed with a flash-free ID and may be reused if not over tightened. Graphite is ideal for glass-to-metal connections because it does not stick to glass. They used for various capillary columns.
Vespel/Graphite ferrules are made from 40% graphite and 60% high temperature DuPont polyimide, withstands 400 °C. They can be reused because they do not seize onto the column, ideal for GC/MS interface applications. Straight ferrules are used for the same dimension tubing.