Retention gaps, fused silica and UltiMetal™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP8088 CP8008 CP8015 CP8007 CP8090 CP8087 CP8080 CP8009 CP8089 CP8017 CP8018 CP8019 CP8066 CP8068 CP8070
VARICP8088EA 299 EUR
VARICP8088 VARICP8008 VARICP8015 VARICP8007 VARICP8090 VARICP8087 VARICP8080 VARICP8009 VARICP8089 VARICP8017 VARICP8018 VARICP8019 VARICP8066 VARICP8068 VARICP8070
Retention gaps, fused silica and UltiMetal™
Slangen Peristaltische pompslangen
The fused silica retention gaps has three different polarities, uncoated for universal applicability. Dimensions equal to 0,53 mm int. Ø fused silica provide easy installation.
  • Easy to install
  • Single continuous length with leak free connections
These fused silica retention gaps with universal CP-Quick-Seal connectors that fit all capillary column sizes. UltiMetal™ retention gaps are complete with coupled high temperature glass-lined connector for ease-of-use.