LaserTab laser printable labels
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
089406 029691 029835
BRDY089406EA 120.98 EUR
BRDY089406 BRDY029691 BRDY029835
LaserTab laser printable labels
Etiketten
Laboratory labels are compatible with office laser printers. The high-performance lab polyester labels feature a low profile to prevent vials from jamming in centrifuges and racks and they can be used in liquid nitrogen to –196 °C (–321 °F) and in autoclaves up to 121 °C (250 °F).