Manual 2-position switching valves, Rheodyne®
Leverancier: IDEX RHEODYNE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7000 7000L
554-2056EA 1330 EUR
554-2056 554-2058
Manual 2-position switching valves, Rheodyne®
Ventielen Regelventielen
Applicable for sample injection, column switching and sample enrichment applications.