Tubular string mops, PharmaPly™ PolyCellulose™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PPM4-MM15 PPM4-MM15IR
MICRPPM4-MM15EA 29.4 EUR
MICRPPM4-MM15 MICRPPM4-MM15IR
Tubular string mops, PharmaPly™ PolyCellulose™
Moppen
An extremely lightweight, tubular string mop that is ideal for covering large surface areas quickly and efficiently in controlled environments, pharmaceutical support areas, hospitals, and laboratories where stringent cleaning routines are imperative.
  • Extremely lightweight, low-linting and super absorbent
  • PolyCellulose strips — 55% polyester, 45% cellulose
  • Twisted strands for an even layer of liquid, without excessive wetting
  • Ideal for large production areas and round-the-clock cleaning cycles
  • Fully saturated mop head is one half the weight of competitive products
  • Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
  • Available gamma irradiated for aseptic environments
  • Compatible with alcohol and strong disinfectants
  • Designed for single use
The Micronova PharmaPly™ PolyCellulose™ Tubular String Mop, when fully saturated, is half the weight of competitive products, making it ergonomically ideal for large production areas and round-the-clock cleaning cycles. The twisted polyester/cellulose strands allow for an even coating, without excessive wetting.
Bestelinformatie: Components are ordered separately.