Hot air sterilisers, large
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E998.1 E999.1
ROTHE998.1EA 470 EUR
ROTHE998.1 ROTHE999.1
Hot air sterilisers, large
Sterilisatoren Hetelucht sterilisatoren
For dry- sterilising of instruments.
  • Glass balls are included
  • Warm-up period 30 minutes
Sterilisation time: 10 to 30 seconds. Units are fitted with glass balls and can reach a temperature of 240 °C.