Volumetric flasks, class A, blue or white markings
Leverancier: Carl Roth
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Rotilabo®
ROTHY292.1EA 19.6 EUR
ROTHY292.1 ROTHY293.1 ROTHY290.1 ROTHY296.1 ROTHY297.1 ROTHY294.1 ROTHY275.1 ROTHY274.1 ROTHY295.1 ROTHY277.1 ROTHY276.1 ROTHY298.1 ROTHY279.1 ROTHY278.1 ROTHY299.1 ROTHY300.1 ROTHY301.1 ROTHY280.1 ROTHY282.1 ROTHY281.1 ROTHY285.1 ROTHY286.1 ROTHY283.1 ROTHY284.1 ROTHY287.1 ROTHY288.1 ROTHY302.1
Volumetric flasks, class A, blue or white markings
Kolven Maatkolven
Borosilicate glass, grade 3.3, with HDPE stopper.
  • Conformity certified with dated batch identification
  • With batch certificate
  • Adjusted to IN
Certificaten: According to DIN EN 12664, ISO 1042