Containers, wide mouth, Rotilabo®
Leverancier: Carl Roth
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ROTHH326.1EA 29.2 EUR
ROTHH326.1 ROTHH314.1 ROTHH315.1 ROTHH325.1 ROTHH323.1 ROTHH312.1 ROTHH324.1 ROTHH313.1 ROTHH318.1 ROTHH319.1 ROTHH329.1 ROTHH328.1 ROTHH316.1 ROTHH317.1 ROTHH327.1 ROTHH321.1 ROTHH332.1 ROTHH322.1 ROTHH331.1 ROTHH320.1 ROTHH330.1
Containers, wide mouth, Rotilabo®
Monsternamepotjes Containers voor algemeen gebruik
Containers are made of hard PVC, clear or amber or clear PETG.
  • Screw-on cap made of PP with PE-foam insert
  • Wide neck enables easy filling
  • Suitable for foodstuffs
Square containers with screw-on cap for packing and storing liquids and solids.