Warning signs, Sekuroka®
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0220.1 H487.1 H498.1 0219.1 0458.1
ROTH0220.1EA 0 EUR
ROTH0220.1 ROTHH487.1 ROTHH498.1 ROTH0219.1 ROTH0458.1
Warning signs, Sekuroka®
Pictogrammen
Made of PE film with colourfast print. Self- adhesive warning symbol labels.
  • Resistant to water, oil and alkalis
  • Temperature stable from −40 to +80 °C
  • Suitable for indoor use
Certificaten: According to DIN 4844, BGV/A8, ASR A1.3 and practice tested.