Funnels with short stem
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A828.1 0583.1 0584.1 0585.1 0586.1 0587.1 0588.1 0589.1 0594.1
ROTHA828.1EA 5.7 EUR
ROTHA828.1 ROTH0583.1 ROTH0584.1 ROTH0585.1 ROTH0586.1 ROTH0587.1 ROTH0588.1 ROTH0589.1 ROTH0594.1
Funnels with short stem
Trechters Filtertrechters Filtertrechters
Funnels, made of natural coloured HDPE, with smooth surface.
With lifting eye and 4 ventilation ribs (except ROTH0583.1).
Leveringsinformatie: ROTHA828.1 is delivered with sieve Ø 50 mm, mesh size 0,4 mm.