Storage drums, Rotilabo®
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A859.1 A860.1 0489.1 0490.1 0491.1
ROTHA859.1EA 331 EUR
ROTHA859.1 ROTHA860.1 ROTH0489.1 ROTH0490.1 ROTH0491.1
Storage drums, Rotilabo®
Vaten, vaten, trommels
Stackable drum, relatively light and impervious to fluids.
  • Ground and brushed inside and out
  • Drum made of 1,4541 stainless steel
  • Lid with silicone seal and 1,4301 stainless steel tension ring
  • Surfaces in contact with product Ra ≤0,6 µm