Spillage trays, Rotilabo®
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0375.1 0376.1 Y756.1
ROTH0375.1EA 0 EUR
ROTH0375.1 ROTH0376.1 ROTHY756.1
Spillage trays, Rotilabo®
Reservoirs/Schalen/Potten Trays/pannen voor gemorste stoffen
Spilling trays made of chrome-nickel steel 18/8.
  • Suitable for food applications
  • Very good chemical resistance
  • Very good temperature resistance
  • Suitable for food applications