In-line filters, Sure-Guard
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-0611-TI2-3 JR-0611-TI05-3 JR-0611-SS2-3 JR-0611-SS05-3
JOURJR-0611-TI2-3EA 175 EUR
JOURJR-0611-TI2-3 JOURJR-0611-TI05-3 JOURJR-0611-SS2-3 JOURJR-0611-SS05-3
In-line filters, Sure-Guard
Filters Lijnfilters
These disposable in-line filters offer an easy and cost-effective way to protect valuable columns against particles.
  • Filter body made of PEEK
  • Maximum flow rate 30 ml/min
  • Pressure rating <350 bar and with 0,4 mm bore size
  • Easily connected directly to any column with an inlet for ¹/₁₆" ext. Ø tubes and 10-32 threads
Features minimal hold-up volume, disposable filter guard, titanium (biocompatible) and stainless-steel frits, permanently pressed in place and two porosity options.