Vloeibare reinigingsmiddelen voor automatische reiniging van glaswerk, neodisher® FA
Leverancier: Dr. Weigert
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
410126 410133 410130
148-3138EA 275 EUR
148-3138 148-3142 148-0162
Vloeibare reinigingsmiddelen voor automatische reiniging van glaswerk, neodisher® FA
Reinigingsmiddelen Detergenten en oppervlaktereinigers
neodisher® FA.
  • Alkaline detergent for the automated reprocessing of thermostable and thermolabile instruments
  • Components: 15 to 30% phosphates
  • Dosage: 2 to 4 ml/L
Components: 15 - 30% phosphates, Disodium and potasium metasilicate.
Dosage: 2 - 4 ml/l
Alkaline liquid concentrate for universal applications Free of phosphates, surfactants and oxidising agents.
Components: 5 - 15% polycarboxylates, NTA (nitrilotriacetic acid).
Opgelet: Strong exothermic reaction with acids. Corrodes aluminium.More Product Information