Poedervormige reinigingsmiddelen voor automatische reiniging van glaswerk, neodisher® LaboClean A8
Leverancier: Dr. Weigert
Ratings:
Avg. Ratings: 3.0 (1 Review)
Total Ratings: 1
Avg. Ratings: 3.0 out of 5
Was this review helpful? Yes No
Yes: 2 No: 0 
agroalimentaire / technicienne qualite / chateauneuf de gadagne
Yes, I recommend
très bon produit de lavage , dommage qu'il n'existe pas en 5kg , système ouverture à l'intérieur pas très pratique sachant qu'il faut le refermer à chaque fois
410276 410266 410287
148-3112EA 138 EUR
148-3112 148-3113 141-0938
Poedervormige reinigingsmiddelen voor automatische reiniging van glaswerk, neodisher® LaboClean A8
Reinigingsmiddelen Detergenten en oppervlaktereinigers
neodisher® LaboClean A8.
  • Alkaline powder for universal applications, free of surfactants
  • Components: < 5% chlorine based bleaching agents; 15 to 30% phosphates; 1 to 5% dichloroisocyanuric acid sodium salt
  • Dosage: 2 to 5 g/L
Components: \< 5% chlorine based bleaching agents; 15 - 30% phosphates; 1 - 5% dichloroisocyanuric acid sodium salt.
Dosage: 2 - 5 g/l
Mildly alkaline powder especially suitable for removal of residues of culture media, tissues, blood, proteins and organic dyes. Free from surfactants.
Components: \< 5% chlorinated bleaching agents (organic chlorine carriers); > 30% phosphates; 1 - 5% dichloroisocyanuric acid sodium salt.
Opgelet: Most of the materials are corrosive or irritant and care must be taken with the choice of materials for cleaning aluminium and titanium items. Please read the material safety data sheet before use.