Magnetic stirring bars, cylindrical, PTFE-encapsulated, Rotilabo®
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2152.1 2153.1 2153.2 0955.1 0955.2 0973.1 0973.2 0989.1 0994.1 0994.2 0999.1 1204.1 1237.1 1292.1 1292.2 1306.1 1306.2 1309.2 1482.1 1482.2 1484.1 1486.1 1486.2 1487.1 1487.2
ROTH2152.1EA 0 EUR
ROTH2152.1 ROTH2153.1 ROTH2153.2 ROTH0955.1 ROTH0955.2 ROTH0973.1 ROTH0973.2 ROTH0989.1 ROTH0994.1 ROTH0994.2 ROTH0999.1 ROTH1204.1 ROTH1237.1 ROTH1292.1 ROTH1292.2 ROTH1306.1 ROTH1306.2 ROTH1309.2 ROTH1482.1 ROTH1482.2 ROTH1484.1 ROTH1486.1 ROTH1486.2 ROTH1487.1 ROTH1487.2
Magnetic stirring bars, cylindrical, PTFE-encapsulated, Rotilabo®
Roerders Magnetische roerstaven
These PTFE-encapsulated micro magnetic stirrer bars are ideal for very small vessels.