Weighing bottles, glass, Rotilabo®
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1367.1 1368.1 1369.1
ROTH1367.1EA 0 EUR
ROTH1367.1 ROTH1368.1 ROTH1369.1
Weighing bottles, glass, Rotilabo®
Flessen Weegflessen
Made of borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
Tall, with ground glass joint stoppers.