Urine collection container with or without snap seals
Leverancier: BOETTGER PAUL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06.044.0002 06.023.0000 06.034.0001 06.034.1101 06.044.1002 06.034.1001 06.013.0100
BOET06.044.0002EA 69.7 EUR
BOET06.044.0002 BOET06.023.0000 BOET06.034.0001 BOET06.034.1101 BOET06.044.1002 BOET06.034.1001 BOET06.013.0100
Urine collection container with or without snap seals
Diagnostische systemen Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP or LDPE, translucent.
  • Resistant to pressure and shatterproof
  • Optimal visibility of content, graduated
  • Fluid tight lid, easy to close
  • Frosted surface on cups and lids for identification marking
Ideal for collecting and storage of urine samples.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Order Now
Specificaties
Environmentally Preferable