m-ColiBlue24® broth
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2608420 2608450 2608442
HACH2608420EA 81.6 EUR
HACH2608420 HACH2608450 HACH2608442
m-ColiBlue24® broth
Media voor microbiologie Geprepareerde media voor microbiologie
In 24 hours, Hach's m-ColiBlue24® broth give the simultaneous results for total coliforms and E. coli. Hach's ready-to-use media eliminates measuring, mixing, and autoclaving steps necessary to prepare dehydrated media. Glass ampules, plastic ampules, 100 ml bottles of m-ColiBlue24® and agar plates, afford maximum shelf life and ease of use for pre-measured and prepared media. Enumerate total coliforms and E. coli on one petri dish. Differentiate colonies easily - red and blue indicate total coliforms and blue specifies E. coli. Get optimal recovery of stressed and injured organisms. Use m-ColiBlue24® broth to accurately monitor and evaluate total coliform and E. coli in drinking water; wastewater; bottled water; beverages; surface, ground, and well water; and ultrapure, chemical processing and pharmaceutical processing waters. With ready-to-use m-ColiBlue24® Broth, you can:
  • Simultaneously detect total coliforms and E. coli in just 24 hours
  • Easily read and interpret results
  • Save money by eliminating special equipment and UV lamps
  • Minimize background growth of non-coliform bacteria, reduce false positives and false negatives, plus get superior sensitivity down to 1 CFU/100 ml
  • Conduct emergency testing and routine monitoring
m-ColiBlue24® should only be used with pads manufactured by the following companies/brands: Pall Corporation (e.g., sterile petri dishes with absorbent pads), Private Label/OEM products manufactured by Pall Gelman, Sartorius (cellulose and glass fibre pads), and Sartorius BioSart products with the integrated Petri dish. No other pad is compatible with the product, and the use of any pad other than those recommended may result in unreliable, inaccurate data.