Conical tubing adapters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-794 P-799
554-3201EA 55.8 EUR
554-3201 UPCHP-799
Conical tubing adapters, Upchurch Scientific®
Fittingen en connectoren
PEEK.
  • Direct connect ¹/₁₆ and ¹/₈" ext. Ø rigid and semi-rigid tubing to peristaltic tubing
  • Accept 0,020 to ¹/₈" (0,50 to 3,2 mm) int. Ø peristaltic tubing
  • Biocompatible flowpath with excellent chemical compatibility
Upchurch Scientific® Conical adapters provide a reliable connection between rigid/semi-rigid tubing and peristaltic-type flexible tubing, such as Tygon® and PharMed®. These adapters are composed of a PEEK polymer female nut, our Super Flangeless™ ferrule system and a Tefzel® (ETFE) or PEEK conical adapter body. The narrow coned end of theMore Product Information