m-Endo broth
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2373520 2373550
HACH2373520EA 44.9 EUR
HACH2373520 HACH2373550
m-Endo broth
Media voor microbiologie Geprepareerde media voor microbiologie
m-Endo broth detects total coliform bacteria (colonies with a greenish-gold sheen). For potable water samples, confirm with Lauryl Tryptose and brilliant green bile broth tubes. Samples that test positive for total coliform bacteria should be further analysed for the presence of faecal coliform or E. coli using EC Medium or EC Medium with MUG respectively.
  • Save valuable time and money using prepared media for membrane filtration methods