Soxhlet extractor head with drain stopcock
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05400143 05400137 05400131 05400149 05400158
533-2149EA 146 EUR
533-2149 LENZ05400137 LENZ05400131 LENZ05400149 LENZ05400158
Soxhlet extractor head with drain stopcock
Extractors Soxhlet-extractors en Soxhlet-type extractors
DURAN®, borosilicate glass, extractor head only.
For the extraction from solids with solvents. The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The drain allows for sampling (after temporary cooling) of the extract to monitor progress of the extraction and also for almost complete recovery of the extract and solvent at the end of the process.